missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSD1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | FSD1L |
---|---|
Concentration | 0.41 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16870625
|
Proteintech
21032-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16860625
|
Proteintech
21032-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FSD1L is a protein coding gene.Specifications
FSD1L | |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q8BYN5, Q9BXM9 | |
313208, 319636, 83856 | |
FSD1L Fusion Protein Ag15345 | |
Primary | |
-20°C | |
FSD1L |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CCDC10, CSDUFD1, FSD1 like protein, FSD1 N terminal like protein, FSD1CL, FSD1L, FSD1NL, MIR1 | |
Fsd1l | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |