missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wesbart™ Freezer Storage Tray
Designed to be placed within a horizontal/ vertical cold store or be free standing
Brand: Wesbart™ WB1247
Code : NEW
Additional Details : Weight : 1.50000kg
Description
Specifications
Plain Storage Tray | |
540 x 136 x 34 mm | |
136 mm |
Specify Anodised Aluminum or Stainless Steel | |
540 mm | |
34 mm |