missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fraction V Bovine Albumin, pH 7, Thermo Scientific Chemicals
Heat-Shock fractionated
£58.09 - £721.38
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15835068
|
Thermo Scientific
J10857.18 |
50 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15845068
|
Thermo Scientific
J10857.22 |
100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15833732
|
Thermo Scientific
J10857.36 |
500 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15875248
|
Thermo Scientific
J10857.A1 |
1 kg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Source: Bovine blood
- Form: Lyophilized powder
- Assay (electrophoresis): ≥98.0%
- Protein: ≥96.0%
- pH (2%, H2O): 6.5 - 7.5
- Loss On Drying: ≤5.0%
- Residue on Ignition: ≤2.0%
- CAS: 9048-46-8
RUO – Research Use Only