missing translation for 'onlineSavingsMsg'
Learn More

FMOC-O-tert-Butyl-L-tyrosine, 98%, Thermo Scientific Chemicals

£32.30 - £65.50

Chemical Identifiers

CAS 71989-38-3
Molecular Formula C28H29NO5
Molecular Weight (g/mol) 459.54
MDL Number MFCD00037129
InChI Key JAUKCFULLJFBFN-KSYWNVGFNA-N
Synonym fmoc-tyr tbu-oh, fmoc-o-tert-butyl-l-tyrosine, fmoc-tyr but-oh, fmoc-tyr t-bu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, fmoc-tyr but, n-fmoc-o-tert-butyl-l-tyrosine, nalpha-fmoc-o-tert-butyl-l-tyrosine, n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine, 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 10895791
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
SMILES CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11965491
View Documents
Thermo Scientific Acros
439090100
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11995491
View Documents
Thermo Scientific Acros
439090250
25 g
£65.50
25g
Please to purchase this item. Need a web account? Register with us today!
Currently Unavailable: We have no information when this product will be available
View alternative products
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

71989-38-3
459.54
JAUKCFULLJFBFN-KSYWNVGFNA-N
10895791
CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
C28H29NO5
MFCD00037129
fmoc-tyr tbu-oh, fmoc-o-tert-butyl-l-tyrosine, fmoc-tyr but-oh, fmoc-tyr t-bu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, fmoc-tyr but, n-fmoc-o-tert-butyl-l-tyrosine, nalpha-fmoc-o-tert-butyl-l-tyrosine, n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine, 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid

Specifications

71989-38-3
White
97.5% min. (HPLC)
MFCD00037129
fmoc-tyr tbu-oh, fmoc-o-tert-butyl-l-tyrosine, fmoc-tyr but-oh, fmoc-tyr t-bu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, fmoc-tyr but, n-fmoc-o-tert-butyl-l-tyrosine, nalpha-fmoc-o-tert-butyl-l-tyrosine, n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine, 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
459.54
459.55
Powder
150°C to 156°C
Authentic
C28H29NO5
-29.5
JAUKCFULLJFBFN-KSYWNVGFNA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
10895791
98%
FMOC-O-tert-Butyl-L-tyrosine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 276-262-7

Product Identifier
  • FMOC-O-tert-Butyl-L-tyrosine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.