Learn More
Thermo Scientific Chemicals Fluorescein Disodium Salt Dihydrate 90%
£20.90 - £71.96
Chemical Identifiers
CAS | 1264198-62-0 |
---|---|
Molecular Formula | C20H10Na2O5·2H2O |
MDL Number | MFCD00150814 |
Synonym | Acid Yellow 73 dihydrate, Fluorescein, water soluble dihydrate, Uranine dihydrate, C.I. 45350 dihydrate |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10752402
|
Thermo Scientific Chemicals
410641000 |
100 g | Glass bottle |
£20.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10558132
|
Thermo Scientific Chemicals
410645000 |
500 g | Glass bottle |
£71.96
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1264198-62-0 | |
C20H10Na2O5·2H2O | |
MFCD00150814 | |
Acid Yellow 73 dihydrate, Fluorescein, water soluble dihydrate, Uranine dihydrate, C.I. 45350 dihydrate |
Specifications
1264198-62-0 | |
C20H10Na2O5·2H2O | |
19, 222 | |
Acid Yellow 73 dihydrate, Fluorescein, water soluble dihydrate, Uranine dihydrate, C.I. 45350 dihydrate | |
Authentic | |
90% | |
88% min. (on dry substance) | |
Powder |
90% | |
MFCD00150814 | |
14, 4159 | |
Solubility in water: soluble. | |
412.3 | |
Pure | |
Brown to Orange/Red or Brown/Green | |
Fluorescein, disodium salt dihydrate, 90% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 208-253-0
missing translation for 'rtecsNumber' : LM5425000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.