missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Flat belt sling, brown, Simplex, 4000kg capacity, four meter length
Brand: Manutan A340159
This item is not returnable.
View return policy
Description
For use when reeving not required.- Vertical lift SWL : 6000kg
- Sling colour: brown
- Width: 300mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction