missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Flat basket sling, 2000kg capacity, green, 1m length
Brand: Manutan A340266
This item is not returnable.
View return policy
Description
For use when reeving not required. Two-ply webbing and steel end fittings. Can be used in conjunction with spreader beams and lifting frames.- Vertical lift SWL: 2000kg
- Green
- Width: 50mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction