Learn More
Liebherr Refrigerator FKv2643 upright, 4 grid shelves plastics coated mechanic control system 220-240V 50Hz 236L capacity +1°C to +15°C dynamic cooling system, free standing, glazed door Liebherr
Brand:  Liebherr FKV 2643
Refrigerator FKv2643 upright, 4 grid shelves plastics coated mechanic control system 220-240V 50Hz 236L capacity +1°C to +15°C dynamic cooling system, free standing, glazed door Liebherr
Code : 03
Additional Details : Weight : 61.00000kg
Product Code. 15193233