Learn More
Mettler Toledo™ Electrode Arm for FiveEasy™ Plus/FiveEasy Meters
Electrode holder allows you to secure sensor when not in use. For use with FiveEasy Plus/FiveEasy pH or conductivity meters.
Brand: Mettler Toledo™ 30239139
Code : NEW
Additional Details : Weight : 0.50000kg
Includes: Sensor holder and a pole
Description
- Accessory for use with FiveEasy Plus/FiveEasy Meters
- Secure your sensor with this ambidextrous electrode holder
- Attach this on the right or left side of the FiveEasy meter for convenient measurement and to secure sensor when not in use
Specifications
FiveEasy/Plus electrode arm | |
Electrode Arm | |
Sensor holder and a pole | |
FiveEasy Meter, FiveEasy Plus Meter |