Learn More
Fisherbrand™ Shorting Strap for BNC Input
For meters with BNC input jack
Brand: Fisherbrand™
Code : 43
Additional Details : Weight : 0.10000kg
Description
For meters with BNC input jack
- Connects input jacks for testing and calibration of meter
Specifications
Shorting Strap | |
pH | |
Meter with BNC input jack |