missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14590-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is highly similar to the rat NFB42 (neural F Box 42 kDa) protein which is enriched in the nervous system and may play a role in maintaining neurons in a postmitotic state.Specifications
FBXO2 | |
Polyclonal | |
Unconjugated | |
FBXO2 | |
F box only protein 2, F box protein 2, FBG1, Fbs1, FBX2, FBXO2, NFB42 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230904, 26232, 85273 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80UW2, Q9UK22 | |
Fbxo2 | |
FBXO2 Fusion Protein Ag6122 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |