Learn More
Thermo Scientific Chemicals Fast Blue BB base
£41.61 - £90.45
Chemical Identifiers
CAS | 120-00-3 |
---|---|
Molecular Formula | C17H20N2O3 |
Molecular Weight (g/mol) | 300.358 |
MDL Number | MFCD00009091 |
InChI Key | CNXZLZNEIYFZGU-UHFFFAOYSA-N |
Synonym | C.I. 37175; 4'-Amino-2',5'-diethoxybenzanilide |
PubChem CID | 67108 |
IUPAC Name | N-(4-amino-2,5-diethoxyphenyl)benzamide |
SMILES | CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15474829
|
Thermo Scientific Chemicals
J62456.14 |
25 g |
£41.61
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15484829
|
Thermo Scientific Chemicals
J62456.22 |
100 g |
£90.45
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C.I. 37175
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C.I. 37175Chemical Identifiers
120-00-3 | |
300.358 | |
CNXZLZNEIYFZGU-UHFFFAOYSA-N | |
67108 | |
CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2 |
C17H20N2O3 | |
MFCD00009091 | |
C.I. 37175; 4'-Amino-2',5'-diethoxybenzanilide | |
N-(4-amino-2,5-diethoxyphenyl)benzamide |
Specifications
95°C to 105°C | |
C17H20N2O3 | |
3416889 | |
CNXZLZNEIYFZGU-UHFFFAOYSA-N | |
N-(4-amino-2,5-diethoxyphenyl)benzamide | |
67108 | |
Brown to Gray | |
Fast Blue BB base |
120-00-3 | |
MFCD00009091 | |
C.I. 37175; 4'-Amino-2',5'-diethoxybenzanilide | |
CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2 | |
300.358 | |
300.35 | |
Powder |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H300-H315-H319-H335-H500
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 204-363-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only