missing translation for 'onlineSavingsMsg'
Learn More

Fast blue B salt, MP Biomedicals™

Product Code. 11446652
Click to view available options
Quantity:
10 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Quantity 100 g
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Fast Blue B Salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'