Learn More
FAM92B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559398
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TLEKYDLERD LLDFRAKMQG VYGHYDTRLL ANTSPPPSVL QSLASQGTLQ VQLSRANEDP EHPHANHGRF SLCEWVVKGQ PAHCVCGQGG Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 32%.
FAM92B Polyclonal antibody specifically detects FAM92B in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
FAM92B | |
Polyclonal | |
Unconjugated | |
FAM92B | |
1700120B06Rik; Fam92b; family with sequence similarity 92 member B; family with sequence similarity 92, member B; protein FAM92B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339145 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZTR7 | |
FAM92B | |
Recombinant protein corresponding to Human FAM92B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only