missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM86B1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563164
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RRAVLPRSHR VAGRGPAEAG CL Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 45%.
FAM86B1 Polyclonal antibody specifically detects FAM86B1 in Human samples. It is validated for Immunohistochemistry.Specifications
FAM86B1 | |
Polyclonal | |
Unconjugated | |
FAM86B1 | |
FAM86B1; family with sequence similarity 86 member B1; family with sequence similarity 86, member B1; protein FAM86B1; putative protein N-methyltransferase FAM86B1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85002 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N7N1 | |
FAM86B1 | |
Recombinant protein corresponding to Human FAM86B1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only