Learn More
FAM172A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA552924
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QIQQGGPDEK EKTTALKDLL SRIDLDELMK KDEPPLDFPD TLEGFEYAFN EKGQLRHIKT GEPFVFNYRE DLHRWNQKRY EALGEIITKY VYELLEKDCN LKKVSIPVDA TESEPKS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 73%.
FAM172A is a protein coding gene.Specifications
FAM172A | |
Polyclonal | |
Unconjugated | |
FAM172A | |
1110033M05Rik; 2610318O14Rik; 53-E6; 9430037D06Rik; AF064782; C5orf21; Cotranscriptional regulator FAM172A; Fam172a; family with sequence similarity 172 member A; family with sequence similarity 172, member A; LOW QUALITY PROTEIN: protein FAM172A; pEN87; Protein FAM172A; RGD1305526 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83989 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WUF8 | |
FAM172A | |
Recombinant protein corresponding to Human FAM172A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only