missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM107A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550409
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total FAM107A protein.
FAM107A (Family With Sequence Similarity 107 Member A) is a Protein Coding gene. Diseases associated with FAM107A include Neuroblastoma and Brain Cancer. An important paralog of this gene isFAM107B.Specifications
FAM107A | |
Polyclonal | |
Unconjugated | |
FAM107A | |
Actin-associated protein FAM107A; actin-associated protein FAM107A; protein FAM107A; down-regulated in renal cell carcinoma 1; DRR1; FAM107A; family with sequence similarity 107 member A; family with sequence similarity 107, member A; protein FAM107A; Protein TU3A; RGD1306327; Tu3a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11170 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95990 | |
FAM107A | |
Full length fusion protein of Human FAM107A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |