Learn More
FAHD2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA559889
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NYVDHCKEQN VPVPKEPIIF SKFASSIVGP YDEVVLPPQS QEVDWEVELA VVIGKKGKHI KATDAMAHVA GFTVAHDVSA RDW Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 84%.
May have hydrolase activity.Specifications
FAHD2A | |
Polyclonal | |
Unconjugated | |
FAHD2A | |
1500003K14Rik; B430104H02Rik; CGI-105; CGI-105 protein; Fahd2; FAHD2A; fumarylacetoacetate hydrolase domain containing 1; fumarylacetoacetate hydrolase domain containing 2A; fumarylacetoacetate hydrolase domain-containing protein 2; Fumarylacetoacetate hydrolase domain-containing protein 2A; LOC100062565; LOC100731203; RGD1563674 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51011 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96GK7 | |
FAHD2A | |
Recombinant protein corresponding to Human FAHD2A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only