missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14906-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an adaptor molecule that interacts with various cell surface receptors and mediates cell apoptotic signals. Through its C-terminal death domain, this protein can be recruited by TNFRSF6/Fas-receptor, tumor necrosis factor receptor, TNFRSF25, and TNFSF10/TRAIL-receptor, and thus it participates in the death signaling initiated by these receptors. Interaction of this protein with the receptors unmasks the N-terminal effector domain of this protein, which allows it to recruit caspase-8, and thereby activate the cysteine protease cascade. Knockout studies in mice also suggest the importance of this protein in early T cell development.Specifications
FADD | |
Polyclonal | |
Unconjugated | |
FADD | |
FADD, GIG3, MORT1, Protein FADD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14082, 266610, 8772 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13158, Q61160 | |
FADD | |
FADD Fusion Protein Ag6701 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |