Learn More
BINDER™ Factory Calibration Certificate
For Use With: BINDER units C 170
Brand:  BINDER™ 8012-0228
Product Code. 15601696
  • £497.00 / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Temperature measurement in center of chamber
  • CO2 measurement performed using test gas at 37°C and 5% CO2
Specifications
Specifications
Calibration certificate, temperature and CO2 - for temperature and CO2, temperature measurement in center of chamber, CO2 measurement performed using test gas at 37°C and 5 % CO2
BINDER Chambers Series CB-S
Calibration Certificate
Temperature and Carbon Dioxide