missing translation for 'onlineSavingsMsg'
Learn More

Factor VIII Related Antigen/von Willebrand Factor Ab-1 Rabbit Polyclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

Brand:  Epredia RB-281-A1

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12653207

  • £154.00 / 500µL
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Factor VIII related antigen or von Willebrand factor is a multimeric glycoprotein. It has functional binding domains to platelet glycoprotein Ib, glycoprotein IIb/IIIa, collagen and heparin. von Willebrand factor is synthesized by endothelial cells and stored in the Weibel-Palade granules. It mediates platelet adhesion to injured vessel walls and serves as a carrier and stabilizer for coagulation factor VIII. von Willebrand factor is one of the most useful markers to identify endothelial (or megakaryocytic) lineage of neoplasms. As not all endothelial cells synthesize/store this molecule, about 30% of tumors of vascular origin fail to stain for factor VIII related antigen, regardless of whether they are benign or malignant. Staining for factor VIII related antigen has also been used to measure angiogenesis an indicator of tumor recurrence.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified factor VIII related antigen/von Willebrand factor

Molecular Weight: 270 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
Specifications

Specifications

Factor VIII Related Antigen/von Willebrand Factor Ab-1
Polyclonal
Rabbit
500 μL
Hematopoietic
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
Purified factor VIII related antigen/von Willebrand factor
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers