Learn More
HAP40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA561382
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LLQPPPAKLL PEHAQTLEKY SWEAFDSHGQ ESSGQLPEEL FLLLQSLVMA THEKDTEAIK SLQVEMWPLL TAEQNHLLHL VLQETISPSG QGV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.
Huntington's disease is caused by an expanded CAG trinucleotide repeat coding for a polyglutamine stretch within the Huntington protein. Huntington co-purifies with a single novel 40 kDa protein designated HAP40. Recombinant HAP40 is cytoplasmic in the presence of Huntington but is actively targeted to the nucleus in the absence of Huntington. These observations suggest that HAP40 °Contributes to the function of normal Huntington and is a candidate for involvement in the aberrant nuclear localization of mutant Huntington found in degenerating neurons in Huntington's disease.Specifications
HAP40 | |
Polyclonal | |
Unconjugated | |
F8a1 | |
40-kDa huntingtin-associated protein; AI852759; coagulation factor VIII-associated (intronic transcript) 1; coagulation factor VIII-associated 1; cpG island protein; DXS522E; DXUcsf1; F8A; F8A1; F8A2; F8A3; factor 8-associated gene A; Factor VIII associated protein; factor VIII intron 22 protein; HAP40; huntingtin-associated protein 40; RP23-114O19.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
474383, 474384, 8263 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23610 | |
F8a1, F8a2, F8a3 | |
Recombinant protein corresponding to Human HAP40 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only