missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One F-Bottom 96-well Assay Microplates

Features a chimney-like arrangement of the wells that allows each well to stand on its own. Greiner Bio One™ F-Bottom 96-Well Assay Microplates are constructed of polystyrene, a highly clear polymer with excellent optical properties making it ideal for precise optical measurements. Polystyrene is also characterized by its ability to bind biomolecules, such as proteins and is suitable for work with cell cultures.

Brand:  Greiner Bio-One 655076

 View more versions of this product

Product Code. 10033211

  • £110.00 / Pack of 40

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Footprint compatible with automated systems
  • Can be traced to production through defined LOT number system
  • Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
  • Manufactured from raw materials tested for leachables
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIN ISO 9001
Standard
34 mm2
Black
96
Non-sterile
392 μL
Yes
4800 x g (swinging bucket rotor)
Yes
Black
5 mm
RNase-/D-Nase, human DNA free
Without Lid
8 x 12
Polystyrene
25 to 34 μL
FLUOTRAC 200
Flat Bottom (F-well)
-20°C to +60°C
40/Cs.
-20°C to +60°C
11.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers