Learn More
EVOS™ EVOS™ Vessel Holder, KOVA™ Glasstic™ slide 10
Allows user to utilize different types of vessels for imaging
Brand:  EVOS™ AMEPVH017
Product Code. 11907335
  • £401.00 / Each
Explore more special offers
Description
Description
  • Fits microscope slide, cell culture flask or dish, or micro-well plate to stage of EVOS imaging systems for increased precision in sample alignment
  • Accommodates one KOVA Glasstic Slide 10
  • Easy-to-change vessel holders and stage plates available for this instrument platform
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vessel Holder
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
1 KOVA™ Glasstic™ Slide 10
EVOS
10 Slides
1 Holder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
EVOS™ EVOS™ Vessel Holder, KOVA™ Glasstic™ slide 10 > KOVA Glasstic Slide 10