missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibertini Elettronica™ EU-C Atex Technical Balances
Designed for use in hazardous environments with the risk of explosion, these scales are classified as ZONE 2. Gibertini Elettronica™ EU-C Atex Technical Balances are built with method of protection nAL for gas group IIC and temperature class T4 (135°C) according to regulation EEC 94/9.
POA - £2055.00
Specifications
Certifications/Compliance | Classified as ZONE 2. Built with method of protect |
---|---|
For Use With (Application) | Designed for use in hazardous environments with th |
Material | Metallic Body with Epoxide Protection |
Temperature Range (Metric) | 10°C to 40°C |
Type | Technical Balance |
Product Code | Brand | Model | Capacity | Readability | Measurement Time | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Capacity | Readability | Measurement Time | Price | Quantity & Availability | ||
15593876
|
Gibertini Elettronica™
EU-C 10000 PTX |
EU-C 15000 X | 15 kg | 0.5 g | 2 sec. |
£2055.00
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15583876
|
Gibertini Elettronica™
EU-C 7500 DRX |
EU-C 10000PTX | 10.2 kg | 0.1 g | 2 sec. |
£1695.00
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15533886
|
Gibertini Elettronica™
EU-C7500 PTX |
EU-C 7500 PTX | 7.5 kg | 0.1 g | 2 sec. |
£1162.88
Each |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15503886
|
Gibertini Elettronica™
EU-C 2002 X |
EU-C 2002 X | ≥2 kg | 0.01 g | 2.5 sec. |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15513886
|
Gibertini Elettronica™
EU-C 4002 X |
EU-C 4200 X | 4.2 kg | 0.01 g | 2.5 sec. |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15307796
|
Gibertini Elettronica™
EU-C 7500 PQX |
EU-C 7500PQX | 7.5 kg | 0.1 g | 2 sec. |
£1695.00
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15523886
|
Gibertini Elettronica™
EU-C7500 PTX |
EU-C 7500 DRX | 950, 7500 g | 0.01, 0.1 g | 1 sec. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- LCD display with small-size decimal digits, for easier and more immediate reading
- Waterproof and acid resistant membrane keyboard. Easy to use with the ON/OFF, TARE, MODE and PRINT keys (RANGE/PRINT for DR model)
- Indication of the reached stable weight
- Bar-graph indicator of dosage and remaining capacity of the balance
- Parameters configurable by menu: reading in g (grams), lb (pound), oz (ounce), ct (carats), pcs (pieces), % (percentage)
- Automatic end of range calibration (with external mass supplied on request)
- Memory of the current weight in case of power failure
- Enhanced anti-shock system
- Metallic body with epoxide protection
- Load cell nearly unbreakable and without transportation problems
- Data output: I/O RS232 adjustable
- 1, 2 and 5kg masses in class F1 for calibration and special executions available upon request
Specifications
Classified as ZONE 2. Built with method of protect | |
Metallic Body with Epoxide Protection | |
Technical Balance | |
240 VAC |
Designed for use in hazardous environments with th | |
10°C to 40°C | |
240 VAC |