missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, ≥99%, Spectrophotometric Grade, Honeywell Riedel-de Haën™

Product Code. 15602960
Click to view available options
Quantity:
1 L
2 L
4 x 2 L
6 x 1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Spectrophotometric Grade,≥99%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -12°C
Density 1.113g/mL at 25°C
Boiling Point 195°C to 198°C
Quantity 4 x 2 L
Explosion Limit 0.153
Flash Point 111°C (231.8°F)
Ignition Point 752°F
Packaging Glass Bottle
Linear Formula HOCH2CH2OH
Vapor Pressure 0.08mmHg (20°C)
UN Number UN3082
Beilstein 505945
Formula Weight 62.07g/mol
Percent Purity ≥99%
Grade Spectrophotometric
Chemical Name or Material Ethylene Glycol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'