missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate, 97%, Thermo Scientific™

£17.30 - £283.00

Chemical Identifiers

CAS 138907-68-3
Molecular Formula C12H12FN3O2
Molecular Weight (g/mol) 249.25
MDL Number MFCD00173917
InChI Key RPPPCKSHIYWAPW-UHFFFAOYSA-N
Synonym ethyl 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluorophenyl pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluoro-phenyl-1h-pyrazole-4-carboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylic acid ethyl ester, 5-amino-4-ethoxycarbonyl-1-4-fluorophenyl-1h-pyrazole, 1-5-amino-4-ethoxycarbonyl-1h-pyrazol-1-yl-4-fluorobenzene, maybridge1_006170, ethyl5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethylaminofluorophenylpyrazolecarboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxyli
PubChem CID 688689
IUPAC Name ethyl 5-amino-1-(4-fluorophenyl)pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(N)N(N=C1)C1=CC=C(F)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10181272
SDS View Documents
Thermo Scientific Maybridge
RH00119DA
1 g
£17.30
1g
Please to purchase this item. Need a web account? Register with us today!
10190552
SDS View Documents
Thermo Scientific Maybridge
RH00119EA
10 g
£127.00
10g
Please to purchase this item. Need a web account? Register with us today!
10031144
SDS View Documents
Thermo Scientific Maybridge
RH00119EB
25 g
£283.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

138907-68-3
249.25
RPPPCKSHIYWAPW-UHFFFAOYSA-N
688689
CCOC(=O)C1=C(N)N(N=C1)C1=CC=C(F)C=C1
C12H12FN3O2
MFCD00173917
ethyl 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluorophenyl pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluoro-phenyl-1h-pyrazole-4-carboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylic acid ethyl ester, 5-amino-4-ethoxycarbonyl-1-4-fluorophenyl-1h-pyrazole, 1-5-amino-4-ethoxycarbonyl-1h-pyrazol-1-yl-4-fluorobenzene, maybridge1_006170, ethyl5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethylaminofluorophenylpyrazolecarboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxyli
ethyl 5-amino-1-(4-fluorophenyl)pyrazole-4-carboxylate

Specifications

138907-68-3
97%
C12H12FN3O2
ethyl 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluorophenyl pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluoro-phenyl-1h-pyrazole-4-carboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylic acid ethyl ester, 5-amino-4-ethoxycarbonyl-1-4-fluorophenyl-1h-pyrazole, 1-5-amino-4-ethoxycarbonyl-1h-pyrazol-1-yl-4-fluorobenzene, maybridge1_006170, ethyl5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethylaminofluorophenylpyrazolecarboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxyli
CCOC(=O)C1=C(N)N(N=C1)C1=CC=C(F)C=C1
249.25
249.24
ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
97.0
Amber glass bottle
MFCD00173917
RPPPCKSHIYWAPW-UHFFFAOYSA-N
ethyl 5-amino-1-(4-fluorophenyl)pyrazole-4-carboxylate
688689
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.