missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (R)-nipecotate L-tartrate, 98%, Thermo Scientific Chemicals
£15.80 - £282.00
Chemical Identifiers
CAS | 167392-57-6 |
---|---|
Molecular Formula | C12H21NO8 |
Molecular Weight (g/mol) | 307.299 |
MDL Number | MFCD00799538 |
InChI Key | HHPGQKZOPPDLNH-RATVZYDNSA-N |
Synonym | s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester |
PubChem CID | 53249980 |
IUPAC Name | (3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate |
SMILES | CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15472668
|
Thermo Scientific Alfa Aesar
H56226.03 |
1 g | N/A | ||||||
15482668
|
Thermo Scientific Alfa Aesar
H56226.06 |
5 g | N/A | ||||||
15492668
|
Thermo Scientific Alfa Aesar
H56226.14 |
25 g |
£282.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
167392-57-6 | |
307.299 | |
HHPGQKZOPPDLNH-RATVZYDNSA-N | |
53249980 | |
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O |
C12H21NO8 | |
MFCD00799538 | |
s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester | |
(3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate |
Specifications
167392-57-6 | |
C12H21NO8 | |
s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester | |
+10° (c=5 in Water) | |
(3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate | |
53249980 | |
98% |
157°C to 159°C | |
MFCD00799538 | |
HHPGQKZOPPDLNH-RATVZYDNSA-N | |
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O | |
307.299 | |
307.3 | |
Ethyl (R)-nipecotate L-tartrate |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only