missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, ≥99.5%, ACS Reagent, Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
6 x 500 mL
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
Melting Point | -84°C |
Density | 0.902g/mL at 25°C |
Boiling Point | 77.5°C |
Quantity | 4 L |
Explosion Limit | 2.2 to 11.5% (38°F) |
Flash Point | −3°C (26.6°F) |
Ignition Point | 801°F |
Packaging | Glass Bottle |
Linear Formula | CH3COOC2H5 |
Vapor Pressure | 73mmHg (20°C) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'