missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanolamine, Certified AR for Analysis, Fisher Chemical™
53.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
53.26 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
53.26 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
Specifications
Melting Point | 10°C |
Color | Colorless |
pH | 12 |
Boiling Point | 170°C |
Packaging | Amber glass bottle |
Quantity | 500 mL |
Vapor Density | 2.1 |
Formula Weight | 61.08g/mol |
Vapor Pressure | 0.48mmHg at 20°C |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'