Learn More
ERK1/2 Monoclonal Antibody (5AD13MA), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 41-9108-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The 5AD13MA monoclonal antibody recognizes human, mouse, and rat extracellular signal-regulated kinases 1 and 2 (also known as ERK1/2, p44/42, or MAPK3/1). ERK1/2 belong to a family of conserved serine/threonine protein kinases known as mitogen-activated protein kinases (MAPKs) that are involved in many cellular programs such as proliferation, differentiation, motility, and survival. The ERK1/2 signaling pathway can be activated in response to a diverse range of extracellular stimuli including mitogens, growth factors, and cytokines. The primary activators of ERK1/2 are MEK1 and MEK2 which activate p44 and p42 through phosphorylation of activation loop residues Thr202/Tyr204 and Thr185/Tyr187 in ERK1 and ERK2, respectively. Several downstream targets of ERK1/2 have been identified, including p90RSK and the transcription factor Elk-1. ERK1/2 are negatively regulated by MAPK phosphatases as well as by chemical inhibitors of MEK including U0126 and PD98059. The ERK pathway is disrupted in many types of cancer and thus is an important target for diagnosis and treatment. Applications Reported: This 5AD13MA antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry. Applications Tested: This 5AD13MA antibody has been tested by immunocytochemistry of methanol-fixed cells and can be used at less than or equal to 5 μg/mL.
ERK1/2 Monoclonal antibody specifically detects ERK1/2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
ERK1/2 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P21708, P27361, P28482, P63085, P63086, Q63844 | |
MAPK1, MAPK3 | |
Affinity chromatography | |
RUO | |
116590, 26413, 26417, 50689, 5594, 5595 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
5AD13MA | |
eFluor 570 | |
MAPK3 | |
12559; 9030612K14Rik; AA407128; AU018647; BcDNA:RE08694; C78273; CG12559; CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; CG18732; CT34260; CT39192; DERK; D-ERK; DERK-A; Diphospho-ERK; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmErk; DmERKA; DmERK-A; DmMAPK; dpERK; dp-ERK; dpERK1; dpMAPK; drosophila ERK; Dsor2; E(sina)7; EC2-1; EK2-1; enhancer of seven in absentia 7; ERK; Erk MAP kinase; Erk/Map kinase; ERK/rolled; Erk1; Erk-1; erk1 erk2; Erk1/2; ERK1b; ERK2; ERK-2; ERKA; ERK-A; ERT1; ERT2; Esrk1; extracellular signal-regulated kinase; extracellular signal-regulated kinase (ERK2); extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; EY2-2; fi06b09; GroupI | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
For Research Use Only.