missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPRS Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25307-1-AP-20UL
This item is not returnable.
View return policy
Description
EPRS is a member of aminoacyl-tRNA synthetases, a class of enzymes that charge tRNAs with their cognate amino acids. EPRS is a multifunctional aminoacyl-tRNA synthetase that catalyzes the aminoacylation of glutamic acid and proline tRNA species.Specifications
EPRS | |
Polyclonal | |
Unconjugated | |
EPRS | |
EARS, EPRS, GLNS, GLUPRORS, GluRS Prolyl tRNA synthetase, Glutamate tRNA ligase, PARS, PIG32, Proline tRNA ligase, QARS, QPRS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2058 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P07814 | |
EPRS | |
EPRS Fusion Protein Ag18011 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |