missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPAC1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | EPAC1 |
---|---|
Concentration | 540 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16810354
|
Proteintech
12572-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16800354
|
Proteintech
12572-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EPAC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EPAC1. This antibody recognizes human antigen. The EPAC1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
EPAC1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG |
540 μg/mL | |
Polyclonal | |
Rabbit | |
10411 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title