missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreenHTS 384-Well Filter Plates

Use in automated, high throughput applications, including biological assays and sample preparation. MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.

Brand:  Merck MZHVN0W50

Additional Details : Weight : 0.01000kg

Product Code. 11794694

  • £3930.00 / Pack of 50
Estimated Shipment: 27-09-2024
to see stock.
Add to basket

Description

Description

For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.
  • Rigid sidewalls allows full compatibility with automated/robotic systems
  • Unique plate design and individually sealed filter discs prevent cross-talk between wells
  • Opaque plastic lids prevents light transfer between wells during direct scintillation counting
  • Barcoding available
  • Meet ANSI™/SBS standards
Specifications

Specifications

Standard
16 x 24
Opaque
384
0.09 cm2
50
1.28 cm
1.44 cm
Durapore™ PVDF
0.45 μm
Styrene
Non-sterile
Receptor binding assays, protein kinase, enzyme assays
1.28 x 8.55 x 1.44 cm
8.55 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers