Learn More
Merck Millipak™ Disposable Filter Units
Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.
Brand: Merck MPHL04GH2
Additional Details : Weight : 0.02000kg
Description
Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.
Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.
- Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
- Point-of-use filter for high-purity water
- Ideally suited for high value-added applications
- Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
- Presterilized units are gamma irradiated
- Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
- 100% integrity tested to guarantee integrity
Specifications
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | |
Polycarbonate | |
0.45 μm | |
200 cm2 | |
Removal of particles and microorganisms from liquids and gases | |
2/Pk. | |
20 L | |
8.9 cm | |
7.6 cm |
Single-use | |
PVDF | |
Sterile | |
8.5 L/min. at 2.45 bar | |
75 psi at 25°C | |
Filter Unit | |
3.5 in. | |
3 in. |