Learn More
DWK Life Sciences™ DURAN™ YOUTILITY™ GL 45 Bottle Tag
Simple color coding for DURAN™ YOUTILITY™ bottles.
Brand: DWK Life Sciences™ 292432875
29.17 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.30000kg
Description
These colorful GL 45 Bottle Tags offer an elegant solution for the marking of laboratory bottles.
- Available in different colors for marking of laboratory bottles and personalization
- Elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
- Feature a graspable tab that makes them easy to remove with bare or gloved hands
- Chemically robust, heat resistant and totally reusable
- Fully compatible with all the GL 45 threads and therefore highly versatile
Specifications
20 Tags/Pk. | |
Black | |
Color Coding and Identification |
Tag | |
Silicone |