Learn More
DWK Life Sciences DURAN™ Triple-Neck Round Bottom Flask, with standard ground joint, angled side necks
Multi-neck pellets are used for distillation, fraction and reactions of all kinds.
Brand:  DWK Life Sciences 241883605
62.57 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10654172
  • £72.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
For distillation, fraction and reaction, mainly multi-neck flasks (two or three neck flasks) are used to be able to install attachments, columns, thermometers, dropping funnels, boiling capillaries, etc. as required.
  • Tight closure due to standard ground joint
  • angled side necks
  • Depending upon the application, accessories, columns, thermometers, dropping funnels, boiling capillaries, etc. can be fitted
  • The geometry permits very uniform heating
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Specifications
85 mm
Borosilicate Glass (3.3)
1/Pk.
Autoclavable
250 mL
Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles
Center neck NS 24/29