Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, RD
To attach.
Brand: DWK Life Sciences 286262206
This item is not returnable.
View return policy
Description
DURAN™ glass threaded tubes with RD thread differ in slightly different angles of inclination in the thread and different tube diameters from the GL thread and have stronger threads. RD threads are not very common.
- Glass threaded tubes with RD thread have a different pitch angle in the thread and different tube diameters compared to GL threads.
- RD threads are characterized by stronger threads
- Safe and easy sealing and loosening of glass-glass components
- Grease-free working possible
- Glass articles for further processing
Specifications
100/Pk. | |
14 mm | |
1.5 mm |
For attachment to glass apparatuses; Installation in glass apparatus and devices | |
RD 14 |