missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences DURAN™ Protect Laboratory Bottle, Protect coated, Clear, With DIN Thread, Plastic Safetry Coated, Graduated Product Code.: 11609908

DWK Life Sciences DURAN™ Protect Laboratory Bottle, Protect coated, Clear, With DIN Thread, Plastic Safetry Coated, Graduated

Product Code. 11609908
250 mL
Click to view available options
Capacity (Metric):
100 mL
150 mL
250 mL
500 mL
750 mL
1000 mL
2000 mL
3500 mL
5000 mL
This item is not returnable. View return policy

Product Code. 11609908

Brand: DWK Life Sciences 218053655

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Multifunctional glass laboratory bottles with protective safety coating.

DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134°C.

  • The coating provides scratch, leak* and splinter* protection (* only applies to bottles 5000 ml and less)
  • Highly resistance, long service-life coating
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • UV protection up to approx. 380 nm due to external coating
  • Complete with screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print
  • Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Coating Plastic
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Includes With screw cap and pouring ring from PP (blue)
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Thread Size (Metric) GL 45
Autoclavable Not Autoclavable
Diameter (Metric) 70 mm
  • GL 45 Protect bottles

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences DURAN™ Protect Laboratory Bottle, Protect coated, Clear, With DIN Thread, Plastic Safetry Coated, Graduated >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.