missing translation for 'onlineSavingsMsg'
Learn More

DRP1 (C-terminal) Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen DRP1 (C-terminal)
Concentration 0.16 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16810944
View Documents
Proteintech
12957-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16800944
View Documents
Proteintech
12957-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dynamin-related protein 1 (DRP1) is a member of the dynamin superfamily of GTPases. Members of this family have diverse cellular functions including vesicle scission, organelle fission, viral resistance, and intracellular trafficking (reviewed in 1). DRP1 affects mitochondrial morphology and is important in mitochondrial and peroxisomal fission in mammalian cells.
Specifications

Specifications

DRP1 (C-terminal)
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse
O00429, Q8K1M6
10059, 74006
DRP1 (C-terminal) Fusion Protein Ag3644
Primary
-20°C
DNM1L
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
DLP1, DNM1L, DRP1
Dnm1l
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications