missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dotriacontane, 97%, Thermo Scientific Chemicals
£21.11 - £225.12
Chemical Identifiers
CAS | 544-85-4 |
---|---|
Molecular Formula | C32H66 |
Molecular Weight (g/mol) | 450.88 |
MDL Number | MFCD00009411 |
InChI Key | QHMGJGNTMQDRQA-UHFFFAOYSA-N |
Synonym | n-dotriacontane, bicetyl, unii-7ksv90rn23, dicetyl, lacceran, dotriacontane, n, dotriacontane, acmc-1akq8, 4-01-00-00595 beilstein handbook reference, wln: 32h |
PubChem CID | 11008 |
ChEBI | CHEBI:36020 |
IUPAC Name | dotriacontane |
SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10230743
|
Thermo Scientific Acros
117690050 |
5 g | Glass bottle |
£21.11
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10162190
|
Thermo Scientific Acros
117690250 |
25 g | Plastic bottle |
£65.73
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10145203
|
Thermo Scientific Acros
117691000 |
100 g | Plastic bottle |
£225.12
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
544-85-4 | |
450.88 | |
QHMGJGNTMQDRQA-UHFFFAOYSA-N | |
11008 | |
dotriacontane |
C32H66 | |
MFCD00009411 | |
n-dotriacontane, bicetyl, unii-7ksv90rn23, dicetyl, lacceran, dotriacontane, n, dotriacontane, acmc-1akq8, 4-01-00-00595 beilstein handbook reference, wln: 32h | |
CHEBI:36020 | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Specifications
544-85-4 | |
Gray to White | |
247°C | |
97% | |
CH3(CH2)30CH3 | |
n-dotriacontane, bicetyl, unii-7ksv90rn23, dicetyl, lacceran, dotriacontane, n, dotriacontane, acmc-1akq8, 4-01-00-00595 beilstein handbook reference, wln: 32h | |
QHMGJGNTMQDRQA-UHFFFAOYSA-N | |
dotriacontane | |
11008 | |
450.86 | |
Shiny Flakes |
67°C to 72°C | |
467°C | |
Authentic | |
C32H66 | |
MFCD00009411 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,soluble in ethyl ether,very soluble in benzene | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
450.88 | |
CHEBI:36020 | |
97% | |
Dotriacontane, 97% |
Safety and Handling
missing translation for 'einecsNumber' : 208-881-5
missing translation for 'rtecsNumber' : JT2360000
missing translation for 'tsca' : TSCA
RUO – Research Use Only