Learn More
Divinyltetramethyldisiloxane, 97%, Thermo Scientific Chemicals
£22.11 - £292.46
Chemical Identifiers
CAS | 2627-95-4 |
---|---|
Molecular Formula | C8H18OSi2 |
Molecular Weight (g/mol) | 186.40 |
MDL Number | MFCD00014933 |
InChI Key | BITPLIXHRASDQB-UHFFFAOYSA-N |
Synonym | 1,3-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, 1,1'-divinyltetramethyldisiloxane, unii-sj4d71zqf4, bisvinyltetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane |
PubChem CID | 75819 |
IUPAC Name | ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane |
SMILES | C[Si](C)(O[Si](C)(C)C=C)C=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10736592
|
Thermo Scientific Acros
249210100 |
10 g | Glass bottle |
£22.11
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10693342
|
Thermo Scientific Acros
249210500 |
50 g | Glass bottle |
£66.23
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10239873
|
Thermo Scientific Acros
249212500 |
250 g | Glass bottle |
£292.46
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2627-95-4 | |
186.40 | |
BITPLIXHRASDQB-UHFFFAOYSA-N | |
75819 | |
C[Si](C)(O[Si](C)(C)C=C)C=C |
C8H18OSi2 | |
MFCD00014933 | |
1,3-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, 1,1'-divinyltetramethyldisiloxane, unii-sj4d71zqf4, bisvinyltetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane | |
ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane |
Specifications
2627-95-4 | |
Colorless | |
139°C | |
Authentic | |
C8H18OSi2 | |
1.4110 to 1.413 | |
0.813 | |
Solubility in water: insoluble | |
C[Si](C)(O[Si](C)(C)C=C)C=C | |
186.40 | |
186.4 | |
Liquid |
-99.7°C | |
0.8130g/mL | |
24°C | |
96% min. (GC) | |
[H2C=CHSi(CH3)2]2O | |
MFCD00014933 | |
1,3-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, 1,1'-divinyltetramethyldisiloxane, unii-sj4d71zqf4, bisvinyltetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane | |
BITPLIXHRASDQB-UHFFFAOYSA-N | |
ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane | |
75819 | |
97% | |
Divinyltetramethyldisiloxane |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 220-099-6
- Divinyltetramethyldisiloxane
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.