missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIO1 Monoclonal Antibody (1E4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524403
This item is not returnable.
View return policy
Description
Peptide Sequence: DRVKRNILAM GEKTGMTRNP HFSHDNWIPT FFSTQYFWFV LKVRWQRLED TTELGGLAPN CPVVRLSGQR CNIWEFMQGN RPLVLNFGSC T
The protein encoded by this gene is a thiol-requiring propylthiouracil-sensitive oxidoreductase. It activates thyroid hormone by converting the prohormone thyroxine (T4) by outer ring deiodination (ORD) to bioactive 3,3',5-triiodothyronine (T3). It also degrades both hormones by inner ring deiodination (IRD). Alternative splicing results in multiple transcript variants encoding different isoforms. Some, but not all, isoforms contain a selenocysteine (Sec) residue encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Additional transcript variants have been described but are not supported by experimental evidence.Specifications
DIO1 | |
Monoclonal | |
Unconjugated | |
Dio1 | |
5DI; D1; deiodinase, iodothyronine type I; deiodinase, iodothyronine, type I; Dio1; DIOI; hDido1; iodothyronine deiodinase type 1; ITDI1; thyroxine 5'-deiodinase; thyroxine deiodinase; thyroxine deiodinase type I (selenoprotein); thyroxine deiodinase, type I; TXDI1; Type 1 DI; type I iodothyronine deiodinase; Type-I 5'-deiodinase | |
Mouse | |
Protein A | |
RUO | |
1733 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
1E4 | |
PBS with no preservative; pH 7.4 | |
P49895 | |
Dio1 | |
DIO1 (NP_000783.2, 35 a.a. ∽ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |