Learn More
Dimethyl cyclohexylmalonate, 95%, Thermo Scientific Chemicals
£48.90 - £158.00
Chemical Identifiers
CAS | 49769-76-8 |
---|---|
Molecular Formula | C11H18O4 |
Molecular Weight (g/mol) | 214.261 |
MDL Number | MFCD05666206 |
InChI Key | CULITIDIVVTWIV-UHFFFAOYSA-N |
Synonym | dimethyl 2-cyclohexylmalonate, dimethyl cyclohexylmalonate, acmc-1asaj, dimethyl cyclohexylpropanedioate, cyclohexylmalonic acid dimethyl ester, cyclohexyl-malonic acid dimethyl ester, 1,3-dimethyl 2-cyclohexylpropanedioate, 2-cyclohexylmalonic acid dimethyl ester, dimethyl 2-cyclohexyl malonate, propanedioic acid, cyclohexyl-, dimethyl ester |
PubChem CID | 2793740 |
IUPAC Name | dimethyl 2-cyclohexylpropanedioate |
SMILES | COC(=O)C(C1CCCCC1)C(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376789
|
Thermo Scientific Alfa Aesar
H26289.03 |
1 g |
£48.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386789
|
Thermo Scientific Alfa Aesar
H26289.06 |
5 g |
£158.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in developing a one-pot, two-step procedure for decarbalkoxylation of malonic esters. The effect of 15C5, other cations, and good nucleophiles was also studied.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in developing a one-pot, two-step procedure for decarbalkoxylation of malonic esters. The effect of 15C5, other cations, and good nucleophiles was also studied.
Solubility
Water soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
49769-76-8 | |
214.261 | |
CULITIDIVVTWIV-UHFFFAOYSA-N | |
2793740 | |
COC(=O)C(C1CCCCC1)C(=O)OC |
C11H18O4 | |
MFCD05666206 | |
dimethyl 2-cyclohexylmalonate, dimethyl cyclohexylmalonate, acmc-1asaj, dimethyl cyclohexylpropanedioate, cyclohexylmalonic acid dimethyl ester, cyclohexyl-malonic acid dimethyl ester, 1,3-dimethyl 2-cyclohexylpropanedioate, 2-cyclohexylmalonic acid dimethyl ester, dimethyl 2-cyclohexyl malonate, propanedioic acid, cyclohexyl-, dimethyl ester | |
dimethyl 2-cyclohexylpropanedioate |
Specifications
49769-76-8 | |
1.4565 | |
dimethyl 2-cyclohexylmalonate, dimethyl cyclohexylmalonate, acmc-1asaj, dimethyl cyclohexylpropanedioate, cyclohexylmalonic acid dimethyl ester, cyclohexyl-malonic acid dimethyl ester, 1,3-dimethyl 2-cyclohexylpropanedioate, 2-cyclohexylmalonic acid dimethyl ester, dimethyl 2-cyclohexyl malonate, propanedioic acid, cyclohexyl-, dimethyl ester | |
CULITIDIVVTWIV-UHFFFAOYSA-N | |
dimethyl 2-cyclohexylpropanedioate | |
2793740 | |
95% |
C11H18O4 | |
MFCD05666206 | |
Water soluble. | |
COC(=O)C(C1CCCCC1)C(=O)OC | |
214.261 | |
214.26 | |
Dimethyl cyclohexylmalonate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.