missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminum hydride, 1M solution in toluene, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Chemical Identifiers

CAS 1191-15-7
MDL Number MFCD00008928
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11369208
SDS View Documents
Thermo Scientific Alfa Aesar
042591.AD
50 mL N/A
N/A
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
15454875
SDS View Documents
Thermo Scientific Alfa Aesar
042591.AE
100 mL N/A
N/A
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
11359208
SDS View Documents
Thermo Scientific Alfa Aesar
042591.AH
200 mL N/A
N/A
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
Specifications

Chemical Identifiers

1191-15-7
MFCD00008928

Specifications

1191-15-7
0.805 g/mL
4°C (40°F)
[(CH3)2CHCH2]2AlH
UN3399
Air and moisture sensitive
142.22
Diisobutylaluminum hydride
Colorless
∼110°C
ChemSeal™ bottles
MFCD00008928
4123663
Miscible with organic solvents.
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H260-H304-H361-H373-H314-H318-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.

P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H250-H260-H304-H314-H335-H336-H361d-H373

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

missing translation for 'einecsNumber' : 214-729-9

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

Product Identifier
  • Diisobutylaluminum hydride, 1M solution in toluene

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 A
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1

Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Toluene, Diisobutylaluminum hydride

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.