Learn More
Gilson™ Digital Dry Bath Block
Click to view available options
Holds:
12 x 15 mL centrifuge tubes
12 x 15 mm or 16 mm test tubes
12 x 5.0 mL centrifuge tubes (17 mm)
20 x 10 mm test tubes or 20 x 2 mL centrifuge tubes
24 x 0.5 mL centrifuge tubes
24 x 1.5 mL centrifuge tubes, conical
24 x 1.5 mL or 2.0 mL centrifuge tubes
48 x 0.2 mL test tubes or 6 PCR strips, 8 tubes each
5 x 50 mL centrifuge tubes
Description
Accessory for Digital Dry Bath Series
Specifications
Specifications
For Use With (Equipment) | Gilson Digital Dry Bath Series |
Holds | 24 x 1.5 mL or 2.0 mL centrifuge tubes |
Type | Block for Digital Dry Bath |