missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylzinc, nominally 15% w/w in hexanes, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Color | Brown to Colorless |
Density | 0.720 g/mL |
Flash Point | −40°C (−40°F) |
Linear Formula | Zn(C2H5)2 |
Quantity | 100 g |
UN Number | UN3394 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
IUPAC Name | diethylzinc |
Formula Weight | 123.49 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction