missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™

£49.35 - £677.00

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
IUPAC Name ethoxyethane
SMILES CCOCC
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10560721
View Documents
Fisher Chemical
D/2400/PB08
500 mL Plastic-coated Glass Bottle
£49.35
500mL
Please to purchase this item. Need a web account? Register with us today!
10396080
View Documents
Fisher Chemical
D/2400/PB17
2.5 L Plastic-coated Glass Bottle
£138.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11389731
View Documents
Fisher Chemical
D/2400/MS21
5 L Aluminum Can
£166.00
5 L
Please to purchase this item. Need a web account? Register with us today!
10131520
View Documents Promotion Details
Fisher Chemical
D/2400/25
25 L Metal Can
£677.00
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

60-29-7
74.12
ethoxyethane
C4H10O
RTZKZFJDLAIYFH-UHFFFAOYSA-N
CCOCC

Specifications

-116°C
Colorless
Liquid
11646
0.2448cp at 20°C
CCOCC
74.12
60-29-7
34.6°C
C4H10O
587mbar at 20°C
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
74.12g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diethyl ether

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3

Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.

SDS
Documents

Documents

Product Certifications