missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072975
Click to view available options
Quantity:
500 mL
2.5 L
This item is not returnable. View return policy
111-42-2
C4H11NO2
2843
This item is not returnable. View return policy
111-42-2
C4H11NO2
2843
Melting Point 27°C
Color Colorless
pH 11
Boiling Point 268°C
Packaging Amber glass bottle
Quantity 500 mL
Formula Weight 105.14g/mol
Vapor Pressure 2 hPa 10
Physical Form Solid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethanolamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'