Learn More
Dichlorophenylsilane, 96%, Thermo Scientific Chemicals
£39.21 - £130.56
Chemical Identifiers
CAS | 1631-84-1 |
---|---|
Molecular Formula | C6H5Cl2Si |
Molecular Weight (g/mol) | 176.09 |
MDL Number | MFCD00039296 |
InChI Key | XNAFLNBULDHNJS-UHFFFAOYSA-N |
Synonym | phenyldichlorosilane, dichlorophenylsilane, dichloro phenyl silane, dichloro phenyl silicon, benzene, dichlorosilyl, phenyl dichlorosilane, phenyl hydrogen dichlorosilane, phenyl-dichlorosilane, phenyldichloro silane, dichloro phenyl silyl |
PubChem CID | 6327324 |
SMILES | Cl[Si](Cl)C1=CC=CC=C1 |
Description
Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from moisture, oxidizing agents. Incompatible materials are bases, moisture and oxidizing agents.
Chemical Identifiers
1631-84-1 | |
176.09 | |
XNAFLNBULDHNJS-UHFFFAOYSA-N | |
6327324 |
C6H5Cl2Si | |
MFCD00039296 | |
phenyldichlorosilane, dichlorophenylsilane, dichloro phenyl silane, dichloro phenyl silicon, benzene, dichlorosilyl, phenyl dichlorosilane, phenyl hydrogen dichlorosilane, phenyl-dichlorosilane, phenyldichloro silane, dichloro phenyl silyl | |
Cl[Si](Cl)C1=CC=CC=C1 |
Specifications
181°C | |
C6H5Cl2Si | |
UN2986 | |
48°C (118°F) | |
Hydrolyzes in water. | |
Cl[Si](Cl)C1=CC=CC=C1 | |
176.09 | |
177.11 | |
1.523 | |
1.212 |
1631-84-1 | |
MFCD00039296 | |
2433865 | |
phenyldichlorosilane, dichlorophenylsilane, dichloro phenyl silane, dichloro phenyl silicon, benzene, dichlorosilyl, phenyl dichlorosilane, phenyl hydrogen dichlorosilane, phenyl-dichlorosilane, phenyldichloro silane, dichloro phenyl silyl | |
XNAFLNBULDHNJS-UHFFFAOYSA-N | |
dichloro(phenyl)silicon | |
6327324 | |
96% | |
Moisture sensitive | |
Dichlorophenylsilane |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 216-635-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only